Learn More
MSC Rubbermaid Cut-to-Length Cotton Dust Mop, White, 40 ft
Catalog No. 502866078
Encompass
This cut-to-length dust mop comes in a 40-foot roll, designed to reduce floor cleaning costs. It is made of untreated, post-industrial 4-ply cotton, offering an economical and recycled material solution. The cut-to-length feature allows customization to fit any standard floor mop frame, featuring a slide-in style backing for secure fit without strings. These floor mops are suitable for commercial and industrial applications.
  • Cut-to-length convenience allows customization to fit any standard mop frame
  • Cut from a continuous 40-foot roll
  • Untreated, cut-end 4-ply cotton yarn provides excellent dust pickup
  • Slide-in style backing for easy, secure fit
  • Made with up to 100% recycled, post-industrial cotton
Catalog No. 50-286-6078 Supplier MSC Supplier No. 09308537
This cut-to-length dust mop comes in a 40-foot roll, designed to reduce floor cleaning costs. It is made of untreated, post-industrial 4-ply cotton, offering an economical and recycled material solution. The cut-to-length feature allows customization to fit any standard floor mop frame, featuring a slide-in style backing for secure fit without strings. These floor mops are suitable for commercial and industrial applications.
  • Cut-to-length convenience allows customization to fit any standard mop frame
  • Cut from a continuous 40-foot roll
  • Untreated, cut-end 4-ply cotton yarn provides excellent dust pickup
  • Slide-in style backing for easy, secure fit
  • Made with up to 100% recycled, post-industrial cotton