Learn More
Essendant Rubbermaid™ Floor Sweeps
Catalog No. 19050657
Change view
Click to view available options
Length (English):
60 in.
Catalog No. Length (English)
19-050-657 60 in.
1 options
Catalog No. 19-050-657 Supplier Essendant Supplier No. RCP6364
Add to Cart
Add to Cart
Floor sweeps feature a durable structural foam block. Medium sweep has sturdy polypropylene fill and may be used indoors or outdoors. Measures 24 in. long (61cm). Silver bristles. Threaded wooden handles are available in either plain lacquered wood or lacquered wood with a metal tip reinforcement; measure 60 in. long (152cm).
Specifications
Product Type Metal-tipped Threaded Broom Handle
For Use With (Application) Used indoors or outdoors, floor sweeps
Length (English) 60 in.
Length (Metric) 152.4 cm
Material Lacquered Wood