Learn More
Beckman Coulter 16mm Diameter Adapter Assembly Tube, Quantity of Four
Catalog No. NC2892486
Encompass_Preferred
Color: PurpleReplacement Rubber Pads: 4Part # 344119Tube Volumes: 12-mLMax Tube Diameter: 16-mmNumber of Places in Adapter: 12Number of Tubes in Rotor: 72Adapter for Tubes in Beckman Coulter Rotors used in High Performance and High Capacity Rotors: JS-4.3Benchtop and Microcentrifuge Rotors: SX4750/4750A
Catalog No. NC2892486 Supplier Beckman Coulter Supplier No. 359162
Color: PurpleReplacement Rubber Pads: 4Part # 344119Tube Volumes: 12-mLMax Tube Diameter: 16-mmNumber of Places in Adapter: 12Number of Tubes in Rotor: 72Adapter for Tubes in Beckman Coulter Rotors used in High Performance and High Capacity Rotors: JS-4.3Benchtop and Microcentrifuge Rotors: SX4750/4750A