Learn More
Creative Biomart Ureaplasma parvum MB antigen, GST-tagged
Catalog No. 502084735
Encompass_Preferred
MB antigen of Ureaplasma parvum serovar fused with GST, is expressed in E. coli, having a Mw of 40kDa, and recognized by the specific IgM anf IgG to Ureaplasma parvum.
Catalog No. 50-208-4735 Supplier Creative Biomart Supplier No. RUB357
MB antigen of Ureaplasma parvum serovar fused with GST, is expressed in E. coli, having a Mw of 40kDa, and recognized by the specific IgM anf IgG to Ureaplasma parvum.