Learn More
Drg International Inc DRG Rubella IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3510R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgG-class antibodies to Rubella Virus in human serum and plasma. The DRG Rubella Virus IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) for Research Use Only. Microtiter wells, as a solid phase, are coated with inactivated K1S grade Rubella Virus antigen (strain HPV-77). Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7586