Learn More
Drg International Inc DRG Rubella IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3511R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgM-class antibodies to Rubella Virus in human serum and plasma. This product is for Research Use Only. The DRG Rubella Virus IgM ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results.
Catalog No. 50-194-7587