Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific Rubella IgM ELISA Test Kit
SDP

Catalog No. 22415203 Shop All Arlington Scientific Products
Change view
Click to view available options
Sample Type:
Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
22-415-203 Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Refer to Package Insert for list of warnings
Catalog No. 22-415-203 Supplier Arlington Scientific Supplier No. 800096AM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Refer to Package Insert for list of warnings

For the qualitative and semiquantitative detection of human IgM antibodies to Rubella virus

  • Runs on the Arlington Scientific Infinity Automated EIA Analyzer
  • Runs on most open system EIA Analyzers
  • Can be performed manually with a plate reader
  • All testing materials provided
  • Ready to use reagents
  • Coated microwell strips

Specifications

Content And Storage 2°C to 8°C
CPT Code 86762
CE Marker Yes
Sample Type Serum
For Use With (Application) ELISA Testing
No. of Wells 96
Sample Size 4 μL
Test Time 90 min. incubation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.