Learn More
Bioss Rubella virus
SDP
Catalog No. 5000065609
Encompass_Preferred
Rubella virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65609 Supplier Bioss Supplier No. BSM72392M
Rubella virus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide