Learn More
ATCC Rubella virus; Strain: RA 27/3
Supplier:  ATCC VR1359
Encompass
Rubella virus strain RA27/3 is propagated in MRC-5 cells (ATCC CCL-171). This animal virus was isolated from a human fetus with congenital rubella and it can be used in infectious disease and respiratory disease research.
Catalog No. 50-238-4322
 Restricted Item