Learn More
Abcam RuBi-Glutamate, caged-glutamate compound, 10MG
Catalog No. 502584605
Encompass_Preferred
MW 970.5 Da. Novel caged-glutamate compound based on ruthenium photochemistry. Excited with visible wavelengths and releases glutamate after one- or two-photon excitation. It has high quantum efficiency and can be used at low concentrations. Enables the photoactivation of neuronal dendrites and circuits with visible or two-photon light sources, achieving single cell, or even single spine, precision. Also available in simple stock solutions (ab146718) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4605 Supplier Abcam Supplier No. AB12040810MG
MW 970.5 Da. Novel caged-glutamate compound based on ruthenium photochemistry. Excited with visible wavelengths and releases glutamate after one- or two-photon excitation. It has high quantum efficiency and can be used at low concentrations. Enables the photoactivation of neuronal dendrites and circuits with visible or two-photon light sources, achieving single cell, or even single spine, precision. Also available in simple stock solutions (ab146718) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement