Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RuBi-Glutamate (hexafluorophosphate sodium) | 2417096-44-5 | 82.9% | 970.54 g/mol | C28H32N5O4PRu·2NaPF6 | 5 MG
SDP

Catalog No. 5000262591
Encompass_Preferred

RuBi-Glutamate (hexafluorophosphate sodium) is a ruthenium-based caged glutamate used to release glutamate with millisecond to sub-microsecond kinetics upon light activation, enabling precise spatial and temporal control in neuroscience and optopharmacology applications.

  • Soluble in water up to 20 mM.
  • Releases glutamate rapidly upon visible-light or two-photon excitation.
  • Enables precise spatial and temporal control of glutamatergic signaling.
  • Suitable for neuronal stimulation and uncaging experiments.
  • Store at -20°C to maintain stability.
  • Provided as 5 mg with 82.9% purity.

Catalog No. 50-002-62591 Supplier Medchemexpress LLC Supplier No. HY1318855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RuBi-Glutamate (hexafluorophosphate sodium) is a ruthenium-based caged glutamate used to release glutamate with millisecond to sub-microsecond kinetics upon light activation, enabling precise spatial and temporal control in neuroscience and optopharmacology applications.

  • Soluble in water up to 20 mM.
  • Releases glutamate rapidly upon visible-light or two-photon excitation.
  • Enables precise spatial and temporal control of glutamatergic signaling.
  • Suitable for neuronal stimulation and uncaging experiments.
  • Store at -20°C to maintain stability.
  • Provided as 5 mg with 82.9% purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.