Learn More
Abcam RuBi-Glutamate (mM/ml), caged-glutamate compound, 10MM_ML
Catalog No. 502704509
Encompass_Preferred
MW 970.5 Da. Novel caged-glutamate compound based on ruthenium photochemistry. Excited with visible wavelengths and releases glutamate after one- or two-photon excitation. It has high quantum efficiency and can be used at low concentrations. Enables the photoactivation of neuronal dendrites and circuits with visible or two-photon light sources, achieving single cell, or even single spine, precision.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4509 Supplier Abcam Supplier No. AB14671810MM_ML
MW 970.5 Da. Novel caged-glutamate compound based on ruthenium photochemistry. Excited with visible wavelengths and releases glutamate after one- or two-photon excitation. It has high quantum efficiency and can be used at low concentrations. Enables the photoactivation of neuronal dendrites and circuits with visible or two-photon light sources, achieving single cell, or even single spine, precision.
The product is subject to the following: Abcam Restricted Use Statement