Learn More
Strem, An Ascensus Company CAS# 7789-39-1. 10g. Rubidium bromide (99%-Rb). MFCD00011186
SDP
Catalog No. 5090115150
Encompass
CAS# 7789-39-1. 10g. Rubidium bromide (99%-Rb). MFCD00011186. Molecular Weight: 165.38. Molecular Formula: RbBr. Color/form: white xtl. Strem# 93-3720. http://www.strem.com/catalog/v/93-3720/
Catalog No. 50-901-15150 Supplier Strem, An Ascensus Company Supplier No. 93372010G
CAS# 7789-39-1. 10g. Rubidium bromide (99%-Rb). MFCD00011186. Molecular Weight: 165.38. Molecular Formula: RbBr. Color/form: white xtl. Strem# 93-3720. http://www.strem.com/catalog/v/93-3720/