Learn More
Alkali Scientific Rubidium Chloride, 1 Gram
SDP
Catalog No. 5000661681
Encompass_Preferred
Rubidium Chloride is an inorganic salt used in laboratory research, particularly in analytical chemistry and biochemistry. The 1-gram vial is ideal for studies requiring rubidium ions, which are often used as a tracer in studies of ion transport and cellular function. Rubidium chloride is also used in various applications such as mass spectrometry, electrophysiology, and isotopic labeling. This reagent is particularly useful in the study of potassium ion transport mechanisms, as rubidium ions behave similarly to potassium in biological systems. Rubidium chloride is essential in cellular biology and analytical techniques requiring precise ionic balance.
Catalog No. 50-006-61681 Supplier Alkali Scientific Supplier No. RUB006CP
Rubidium Chloride is an inorganic salt used in laboratory research, particularly in analytical chemistry and biochemistry. The 1-gram vial is ideal for studies requiring rubidium ions, which are often used as a tracer in studies of ion transport and cellular function. Rubidium chloride is also used in various applications such as mass spectrometry, electrophysiology, and isotopic labeling. This reagent is particularly useful in the study of potassium ion transport mechanisms, as rubidium ions behave similarly to potassium in biological systems. Rubidium chloride is essential in cellular biology and analytical techniques requiring precise ionic balance.