Learn More
Alkali Scientific Rubidium Chloride, 100 Grams
SDP
Catalog No. 5000661720
Encompass_Preferred
Rubidium Chloride is an inorganic salt supplied in crystalline form and used as a reagent in various laboratory applications. This 100g packaging is ideal for laboratories requiring rubidium salts in research projects related to ion transport, chromatography, and atomic spectroscopy. Rubidium chloride is also used in specialized studies of atomic absorption, as rubidium ions are closely related to potassium ions, making them useful in physiological and biochemical assays. It is often employed as a reference material in analytical techniques that involve flame emission spectroscopy and ion-exchange chromatography, providing researchers with precise analytical measurements of metal ions.
Catalog No. 50-006-61720 Supplier Alkali Scientific Supplier No. RUB004CP
Rubidium Chloride is an inorganic salt supplied in crystalline form and used as a reagent in various laboratory applications. This 100g packaging is ideal for laboratories requiring rubidium salts in research projects related to ion transport, chromatography, and atomic spectroscopy. Rubidium chloride is also used in specialized studies of atomic absorption, as rubidium ions are closely related to potassium ions, making them useful in physiological and biochemical assays. It is often employed as a reference material in analytical techniques that involve flame emission spectroscopy and ion-exchange chromatography, providing researchers with precise analytical measurements of metal ions.