Learn More
Indofine Chemical Rubidium Chloride, 99%, 5 Gm
SDP
Catalog No. 501006566
Encompass_Preferred
Formula: ClRb, Purity: 99%, MW: 120.92, MP: 715C, Appearance: White crystals, Storage conditions: Store in a cool, dry place
Catalog No. 50-100-6566 Supplier Indofine Chemical Supplier No. BIO347 5 GM
Formula: ClRb, Purity: 99%, MW: 120.92, MP: 715C, Appearance: White crystals, Storage conditions: Store in a cool, dry place