Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium chloride, 99.8% (metals basis)

Catalog No. AA1056318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1056318 50 g
AA1056309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1056318 Supplier Thermo Scientific Chemicals Supplier No. 01056318
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7791-11-9 | ClRb | 120.92 g/mol

Rubidium chloride is the mostly used rubidium compound, and finds use in various fields ranging from electrochemistry to molecular biology. It is an efficient non-invasive biomarker; increases dopamine and norepinephrine levels, and useful for anergic and apathetic depressives. Pharmaceutical compositions have been used as antidepressants in Europe. It is employed in biochemistry to induce cells to take up DNA. This compound increases the competence of the cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+);chloride
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Color White
Physical Form Crystalline
Quantity 50 g
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Assay Percent Range 99.8% (metals basis)
Density 2.800 g/mL
Linear Formula RbCl
Merck Index 14,8284
Solubility Information Soluble in water. Sparingly soluble in methanol.
Formula Weight 120.92
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Rubidium chloride
Show More Show Less
EINECSNumber 232-240-9
RTECSNumber VL8575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.