Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium chloride, 99% (metals basis)

Catalog No. aa1289214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1289214 25 g
AA1289206 5 g
AA1289222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1289214 Supplier Thermo Scientific Chemicals Supplier No. 01289214
Only null left
Add to Cart

Testing for perchloric acid, source of rubidium, catalyst | CAS: 7791-11-9 | ClRb | 120.92 g/mol

It is used in testing for perchloric acid in laboratories, and is a source of rubidium. Rubidium chloride is used as a catalyst and additive in petrol. It has been employed to prepare molecular nanowires for nanoscale devices. Thin layer of rubidium chloride enhance the attributes of organic light-emitting diodes. Rubidium chloride also plays an essential role for enhancement in the memory storage property. It is an excellent non-invasive biomarker, and used in biomedical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 718°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99% (metals basis)
Merck Index 14,8284
Solubility Information Soluble in water; Practically insoluble in alcohol.
IUPAC Name rubidium(1+);chloride
Formula Weight 120.92
Percent Purity 99%
Odor Odorless
Refractive Index 1.493
Sensitivity Hygroscopic
Appearance White crystals
Density 2.8
Assay (metals basis)
Chemical Name or Material Rubidium chloride
Show More Show Less
EINECSNumber 232-240-9
RTECSNumber VL8575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.