Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium Chloride, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1076150025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1076150025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1076150025 Supplier MilliporeSigma Supplier No. 1076150025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity rubidium chloride for regulated analyses and advanced laboratory applications

  • Emsure™ grade rubidium chloride
  • Premium grade for regulated analyses and advanced laboratory applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Boiling Point 1390°C
Melting Point 715°C
pH 7
Physical Form Solid
Quantity 25 g
Assay Percent Range ≥98.0% (argentometric)
Solubility Information Soluble in Water
Formula Weight 120.92. (molar mass)
Grade For Analysis
Concentration or Composition (by Analyte or Components) Assay (argentometric): ≥98.0 %; Sulfate (SO4): ≤0.2 %; Total Nitrogen (N): ≤0.003 %; Al (Aluminium): ≤0.001 %; Cu (Copper): ≤0.0003 %; Fe (Iron): ≤0.0003 %; Mg (Magnesium): ≤0.0005 %; Pb (Lead): ≤0.003 %; Zn (Zinc):≤ 0.0003 %
Manufacturing Origin Germany
Identification Passes Test
Odor Odorless
Packaging Glass bottle
Product Line Emsure™
Appearance White solid
Specific Gravity 2.76
Density 2.76 g/cm³
Chemical Name or Material Rubidium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.