Learn More
Description
- Crystalline powder with an optical grade
- Key Applications: Catalyst, additive, preparation of molecular nanowires, transfection of bacteria, and investigation of ion channel gating and permeability
- UV Visible Absorbance: OD280(40% aq soln)≤0.2
- MP Biomedicals inorganic chemicals are suitable for laboratory applications
Specifications
Specifications
| Boiling Point | 1390°C |
| Melting Point | 715°C |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | Metal min. 99%. |
| Solubility Information | Soluble in Water |
| Formula Weight | 120.9 |
| Grade | Optical |
| Manufacturing Origin | United States |
| Sensitivity | Air, Moisture Sensitive |
| Show More |
Safety and Handling
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.