Learn More
Cayman Chemical RubIgInon D2 2.5mg
SDP
Catalog No. 502991906
Encompass_Preferred
A polyketide; active against S. aureus and E. coli in agar diffusion assays at 64 UG/disc; inhibits growth of HM02, KATO III, HepG2, and MCF-7 cancer cells (GI50s = 0.1, 0.7, <0.1, and 7.5 µmol/L)
Catalog No. 50-299-1906 Supplier Cayman Chemical Supplier No. 314632.5 MG
A polyketide; active against S. aureus and E. coli in agar diffusion assays at 64 UG/disc; inhibits growth of HM02, KATO III, HepG2, and MCF-7 cancer cells (GI50s = 0.1, 0.7, <0.1, and 7.5 µmol/L)