Learn More
AdipoGen Rubiginone A2
SDP
Supplier:  AdipoGen BVT0023M001
Encompass_Preferred
Chemical. CAS 96695-58-8. Formula C20H16O5. MW 336.3. Isolated from Streptomyces sp. strain Gö N1/5. Antibiotic. Antibacterial. Stereoisomer of SNA 8073B which is a prolyl endopeptidase PEP inhibitor. Vincristine-induced cytotoxicity potentiating activity against multi-drug-resistant tumor cells.
Catalog No. 50-256-2146