Learn More
AdipoGen -Rubiginone B2
SDP
Supplier:  AdipoGen BVT0026M001
Encompass_Preferred
Chemical. CAS 130548-10-6. Formula C20H16O4. MW 320.3. Isolated from Streptomyces sp. Angucyclinone antibiotic. Antibacterial and antitumor agent. Potentiates the cytotoxicity of vincristine. Antimalarial agent.
Catalog No. 50-256-2142