Learn More
AdipoGen Rubiginone D2
SDP
Catalog No. 502562144
Encompass_Preferred
Chemical. CAS 274913-71-2. Formula C20H16O6. MW 352.3. Isolated from Streptomyces sp. strain Gö N1/5. Antibiotic. Antibacterial. Antitumor compound.
Catalog No. 50-256-2144 Supplier AdipoGen Supplier No. BVT0024M001
Chemical. CAS 274913-71-2. Formula C20H16O6. MW 352.3. Isolated from Streptomyces sp. strain Gö N1/5. Antibiotic. Antibacterial. Antitumor compound.