Learn More
Electron Microscopy Sciences Rubis Micro Scissors Set – 3 per set
SDP
Catalog No. 50364979
Encompass_Preferred
High quality scissors, manufactured from hardened, anti-acid, stainless, surgical steel (CA) Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist grip scissors offer ergonomically, comfort and control. Rubis Colibri fine scissors, light duty, Classic medium fine scissors, Sauro medium strong scissors, heavy duty.
Catalog No. 50-364-979 Supplier Electron Microscopy Sciences Supplier No. 7818003
High quality scissors, manufactured from hardened, anti-acid, stainless, surgical steel (CA) Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist grip scissors offer ergonomically, comfort and control. Rubis Colibri fine scissors, light duty, Classic medium fine scissors, Sauro medium strong scissors, heavy duty.