Learn More
Electron Microscopy Sciences Rubis Tweezers Style 5AG
SDP
Catalog No. 501901719
Encompass_Preferred
Light weight tweezers have ultra fine presision tips are engineered with a non-slip grip. GRIP. Made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA). Also resistent to hydrogen fluoride (HF) and nitric acid. Melting point 1400 degree C.
OAL: 110 mm
Tip Width: 9 mm
Gap: 1.5 mm
Tip length: 30 mm
Catalog No. 50-190-1719 Supplier Electron Microscopy Sciences Supplier No. 781705AG
Light weight tweezers have ultra fine presision tips are engineered with a non-slip grip. GRIP. Made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA). Also resistent to hydrogen fluoride (HF) and nitric acid. Melting point 1400 degree C.
OAL: 110 mm
Tip Width: 9 mm
Gap: 1.5 mm
Tip length: 30 mm