Learn More
Medchemexpress LLC HY-13744 10mg Medchemexpress, Rubitecan CAS:91421-42-0 Purity:>98%
SDP
Catalog No. 501932686
Encompass_Preferred
Medchemexpress, HY-13744 10mg Rubitecan CAS:91421-42-0 Rubitecan (RFS 2000), a Camptothecin derivative, is an orally active topoisomerase I inhibitor with broad antitumor activity, and induces protein-linked DNA single-strand breaks, thereby blocking DNA and RNA synthesis in dividing cells [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2686 Supplier Medchemexpress LLC Supplier No. HY1374410MG
Add to Cart
Add to Cart
Medchemexpress, HY-13744 10mg Rubitecan CAS:91421-42-0 Rubitecan (RFS 2000), a Camptothecin derivative, is an orally active topoisomerase I inhibitor with broad antitumor activity, and induces protein-linked DNA single-strand breaks, thereby blocking DNA and RNA synthesis in dividing cells [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.