Learn More
Medchemexpress LLC Ruboxistaurin | 169939-94-0 | 99.5% | 468.55 g/mol | C28H28N4O3 | 100 MG
SDP
Catalog No. 5000248839
Encompass_Preferred
Ruboxistaurin is a selective, orally active protein kinase C beta (PKCβ) inhibitor for preclinical research into PKCβ-related signaling and diabetic vascular complications. It is provided as a solid and as a DMSO solution, and is characterized by defined physicochemical properties and high purity to support reproducible experimental work.
  • Selective PKCβ inhibition for targeted signaling studies.
  • High purity (99.5%) supporting reliable experimental results.
  • Well-characterized molecular properties (C28H28N4O3; 468.55 g/mol).
  • Available in multiple formats and pack sizes for flexible experimental needs.
  • CAS registry linkage for unambiguous compound identification.
Catalog No. 50-002-48839 Supplier Medchemexpress LLC Supplier No. HY10195100MG
Ruboxistaurin is a selective, orally active protein kinase C beta (PKCβ) inhibitor for preclinical research into PKCβ-related signaling and diabetic vascular complications. It is provided as a solid and as a DMSO solution, and is characterized by defined physicochemical properties and high purity to support reproducible experimental work.
  • Selective PKCβ inhibition for targeted signaling studies.
  • High purity (99.5%) supporting reliable experimental results.
  • Well-characterized molecular properties (C28H28N4O3; 468.55 g/mol).
  • Available in multiple formats and pack sizes for flexible experimental needs.
  • CAS registry linkage for unambiguous compound identification.