Learn More
Selleck Chemical LLC Ruboxistaurin
SDP
Catalog No. 5000003587
Encompass_Preferred
Ruboxistaurin (LY333531) HCl is a -specific protein kinase C inhibitor It competitively and reversibly inhibits PKC 1 and PKC 2 with IC50 values of 4 7 and 5 9 nM respectively
Catalog No. 50-000-03587 Supplier Selleck Chemical LLC Supplier No. S76635MG
Ruboxistaurin (LY333531) HCl is a -specific protein kinase C inhibitor It competitively and reversibly inhibits PKC 1 and PKC 2 with IC50 values of 4 7 and 5 9 nM respectively