Learn More
Medchemexpress LLC Ruboxistaurin hydrochloride | 169939-93-9 | 99.9% | 505.01 g/mol | C28H29ClN4O3 | 100 MG
SDP
Catalog No. 5000247867
Encompass_Preferred
Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin, a potent, selective inhibitor of protein kinase C beta (PKCβ) supplied as an orange-red solid for research use. It is used to study PKCβ-mediated signaling, diabetic vascular complications, and related pharmacology. Molecular formula C28H29ClN4O3; molecular weight 505.01 g/mol; CAS 169939-93-9.
  • Selective PKCβ inhibition with low-nanomolar potency.
  • Suitable for in vitro and in vivo PKCβ research.
  • High purity (≈99.9%) supports reproducible assays.
  • Orange-red solid, stable for standard storage conditions.
  • Soluble in DMSO to prepare concentrated stock solutions.
Catalog No. 50-002-47867 Supplier Medchemexpress LLC Supplier No. HY10195B100MG
Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin, a potent, selective inhibitor of protein kinase C beta (PKCβ) supplied as an orange-red solid for research use. It is used to study PKCβ-mediated signaling, diabetic vascular complications, and related pharmacology. Molecular formula C28H29ClN4O3; molecular weight 505.01 g/mol; CAS 169939-93-9.
  • Selective PKCβ inhibition with low-nanomolar potency.
  • Suitable for in vitro and in vivo PKCβ research.
  • High purity (≈99.9%) supports reproducible assays.
  • Orange-red solid, stable for standard storage conditions.
  • Soluble in DMSO to prepare concentrated stock solutions.