Learn More
Medchemexpress LLC Ruboxistaurin hydrochloride | 169939-93-9 | MFCD00940226 | 99.9% | 505.01 | C28H29ClN4O3 | 10 MG
SDP
Catalog No. 5000247967
Encompass_Preferred
Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin (LY-333531), a selective and orally active protein kinase C beta (PKCβ) inhibitor used in preclinical research. It inhibits PKCβ isoforms in the low-nanomolar range and is commonly used in studies of diabetic vascular complications and PKC-related signaling.
  • Selective protein kinase C beta (PKCβ) inhibitor with low-nanomolar potency.
  • Hydrochloride salt form for improved solubility in common solvents.
  • High chemical purity suitable for research applications.
  • Available in small research quantities, including 10 mg packaging.
  • Product documentation includes a datasheet and safety data sheet.
Catalog No. 50-002-47967 Supplier Medchemexpress LLC Supplier No. HY10195B10MG
Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin (LY-333531), a selective and orally active protein kinase C beta (PKCβ) inhibitor used in preclinical research. It inhibits PKCβ isoforms in the low-nanomolar range and is commonly used in studies of diabetic vascular complications and PKC-related signaling.
  • Selective protein kinase C beta (PKCβ) inhibitor with low-nanomolar potency.
  • Hydrochloride salt form for improved solubility in common solvents.
  • High chemical purity suitable for research applications.
  • Available in small research quantities, including 10 mg packaging.
  • Product documentation includes a datasheet and safety data sheet.