Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ruboxistaurin hydrochloride | 169939-93-9 | MFCD00940226 | 99.9% | 505.01 g/mol | C28H29ClN4O3 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY10195B1MG

Encompass_Preferred

Ruboxistaurin hydrochloride is the hydrochloride salt of ruboxistaurin (LY333531), a selective ATP-competitive inhibitor of protein kinase C beta (PKCβ) supplied for research use in biochemical, cellular, and in vivo studies probing PKCβ-mediated signaling.

  • Selective PKCβ inhibition with low-nanomolar potency.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO (25 mg/mL) with ultrasonic warming.
  • Storage: sealed at 4°C; in solvent -20°C (1 month) or -80°C (6 months).
  • Supplied in small research pack sizes such as 1 mg.

Catalog No. 50-002-47986


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.