Learn More
Medchemexpress LLC Ruboxistaurin hydrochloride | 169939-93-9 | MFCD00940226 | 99.9% | 505.01 g/mol | C28H29ClN4O3 | 25 MG
SDP
Catalog No. 5000246581
Encompass_Preferred
Ruboxistaurin hydrochloride is an orally active, selective protein kinase C beta (PKCβ) inhibitor used in research to investigate PKCβ-mediated signaling and complications of diabetes. It is a solid, orange to red compound (CAS 169939-93-9) with molecular formula C28H29ClN4O3 and molecular weight 505.01 g/mol. The product is supplied at high purity and includes recommended storage conditions for both solid material and solutions.
  • Selective PKCβ inhibitor with low-nanomolar potency.
  • Exhibits ATP-dependent competitive inhibition of PKCβ.
  • Displays limited activity against other PKC isoforms.
  • Reduces glucose-induced monocyte adherence in vitro.
  • Store sealed at 4°C; in solution, keep at -80°C for long-term storage.
Catalog No. 50-002-46581 Supplier Medchemexpress LLC Supplier No. HY10195B25MG
Ruboxistaurin hydrochloride is an orally active, selective protein kinase C beta (PKCβ) inhibitor used in research to investigate PKCβ-mediated signaling and complications of diabetes. It is a solid, orange to red compound (CAS 169939-93-9) with molecular formula C28H29ClN4O3 and molecular weight 505.01 g/mol. The product is supplied at high purity and includes recommended storage conditions for both solid material and solutions.
  • Selective PKCβ inhibitor with low-nanomolar potency.
  • Exhibits ATP-dependent competitive inhibition of PKCβ.
  • Displays limited activity against other PKC isoforms.
  • Reduces glucose-induced monocyte adherence in vitro.
  • Store sealed at 4°C; in solution, keep at -80°C for long-term storage.