Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ruboxistaurin mesylate | 192050-59-2 | 99.9% | 564.65 g/mol | C29H32N4O6S | 50 MG
SDP

Catalog No. 5000247866
Encompass_Preferred

Ruboxistaurin mesylate is an orally active, selective, ATP-competitive protein kinase C beta (PKCβ) inhibitor used in preclinical research on diabetic complications, retinal disorders, and cardiovascular conditions. Supplied as the mesylate salt, it is an orange-to-red solid with molecular formula C29H32N4O6S and molecular weight 564.65 g/mol.

  • Selective PKCβ inhibition with low nanomolar potency (IC50 ~4.7-5.9 nM).
  • Mesylate salt form for improved stability and handling.
  • High purity (99.87%) suitable for research applications.
  • Orange-to-red solid, convenient for solid-state storage.
  • Molecular formula C29H32N4O6S and molecular weight 564.65 g/mol.

Catalog No. 50-002-47866 Supplier Medchemexpress LLC Supplier No. HY10195A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ruboxistaurin mesylate is an orally active, selective, ATP-competitive protein kinase C beta (PKCβ) inhibitor used in preclinical research on diabetic complications, retinal disorders, and cardiovascular conditions. Supplied as the mesylate salt, it is an orange-to-red solid with molecular formula C29H32N4O6S and molecular weight 564.65 g/mol.

  • Selective PKCβ inhibition with low nanomolar potency (IC50 ~4.7-5.9 nM).
  • Mesylate salt form for improved stability and handling.
  • High purity (99.87%) suitable for research applications.
  • Orange-to-red solid, convenient for solid-state storage.
  • Molecular formula C29H32N4O6S and molecular weight 564.65 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.