Learn More
Medchemexpress LLC Ruboxistaurin mesylate | 192050-59-2 | MFCD09970504 | 99.9% | 564.65 | C29H32N4O6S | 25 MG
SDP
Catalog No. 5000245868
Encompass_Preferred
Ruboxistaurin mesylate is the methanesulfonate salt of ruboxistaurin, a potent, orally active, ATP-competitive inhibitor of protein kinase C beta (PKCβ). It exhibits low-nanomolar potency against PKCβ isoforms and is supplied as a high-purity solid for in vitro and preclinical research into PKCβ-related signaling, diabetic complications, and cardiovascular or ocular disease models.
  • Selective PKCβ inhibition (IC50 ≈ 4.7-5.9 nM).
  • High purity suitable for research use.
  • Mesylate salt form with DMSO solubility for assay preparation.
  • Supplied as a characterized solid with batch documentation.
  • Appropriate for in vitro and preclinical pathway and disease-model studies.
Catalog No. 50-002-45868 Supplier Medchemexpress LLC Supplier No. HY10195A25MG
Ruboxistaurin mesylate is the methanesulfonate salt of ruboxistaurin, a potent, orally active, ATP-competitive inhibitor of protein kinase C beta (PKCβ). It exhibits low-nanomolar potency against PKCβ isoforms and is supplied as a high-purity solid for in vitro and preclinical research into PKCβ-related signaling, diabetic complications, and cardiovascular or ocular disease models.
  • Selective PKCβ inhibition (IC50 ≈ 4.7-5.9 nM).
  • High purity suitable for research use.
  • Mesylate salt form with DMSO solubility for assay preparation.
  • Supplied as a characterized solid with batch documentation.
  • Appropriate for in vitro and preclinical pathway and disease-model studies.