Learn More
AdipoGen Rubpy2-5-ClAPA-PF62
SDP
Supplier:  AdipoGen CDXR0028M100
Encompass_Preferred
Chemical. CAS 204273-42-7. Formula C34H26F12ClN7OP2Ru. MW 975.0679. Synthetic. Reactive probe containing s fluorescent metal-ligand complex for the determination of nucleotide base sequences.
Catalog No. 50-256-0125