Learn More
Abcam Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2, ester of ruthenium, 1MG
Catalog No. 502705335
Encompass_Preferred
MW 1014.66 g/mol, Purity >97%. Activated ester of ruthenium for acylating amino acid side chain amines; can be used for 1D- or 2D-protein gel staining.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5335 Supplier Abcam Supplier No. AB1452851MG
MW 1014.66 g/mol, Purity >97%. Activated ester of ruthenium for acylating amino acid side chain amines; can be used for 1D- or 2D-protein gel staining.
The product is subject to the following: Abcam Restricted Use Statement