Learn More
AdipoGen Rubpy2mcbpy-O-Su-estPF62
SDP
Supplier:  AdipoGen CDXB0032M001
Encompass_Preferred
Chemical. CAS 136724-73-7. Formula C36H29F12N7O4P2Ru. MW 1014.66. Synthetic. Activated ester of ruthenium complex for acylation of amino acid side chain amines. This label is perfectly suitable for 1D- or 2D-protein gel staining. A simple pre-electrophoresis procedure provides a sensitivity better than SYPRO Ruby and a similar dynamic range. In contrast to SYPRO Ruby, this staining exhibits a logarithmic dependency on the amount of protein.
Catalog No. 50-256-1700