Learn More
AdipoGen Rubpy2mcbpyPF62
SDP
Supplier:  AdipoGen CDXB0658M010
Encompass_Preferred
Chemical. CAS 136724-83-9. Formula Rubpy2mCbpy PF62. MW 917.59. Intermediate to produce the activated Rubpy2mcbpy-O-Su-esterPF62 ester, suitable for 1D- or 2D-protein gel staining and for acylation of amino acid side chain amines. Used to derivatize polymers and oligonucleotides and for luminescent applications.
Catalog No. 50-256-1574