Learn More
AdipoGen Rubpy2phen-5-NH2PF62
SDP
Catalog No. 502561573
Encompass_Preferred
Chemical. CAS 84537-86-0. Formula C32H25N7Ru . 2PF6. MW 898.59. Rubpy2phen-5-NH2PF62 is a fluorescent metal-ligand complex probe. It shows high brightness and large Stokes Shift. Spectral Data lambdaex=458nm lambdaem=612nm in 0.1 M Tris pH 7.0. It has been used as starting material for synthesis of long-lifetime luminescent Ru complexes and in the preparation of amino-rutheniumii complex functionalized gold nanoparticles, which could be used as probe in biosensors and bioassays. Rubpy2phen-5-NH2PF62 is has also been used to synthesize fluorescent silica nanoparticles with ultrabrightness and photostability by encapsulating it into silica nanoparticles by a modified reversed microemulsion method. Could be used for the determination of nucleotide base sequences.
Catalog No. 50-256-1573 Supplier AdipoGen Supplier No. CDXB0671M005
Chemical. CAS 84537-86-0. Formula C32H25N7Ru . 2PF6. MW 898.59. Rubpy2phen-5-NH2PF62 is a fluorescent metal-ligand complex probe. It shows high brightness and large Stokes Shift. Spectral Data lambdaex=458nm lambdaem=612nm in 0.1 M Tris pH 7.0. It has been used as starting material for synthesis of long-lifetime luminescent Ru complexes and in the preparation of amino-rutheniumii complex functionalized gold nanoparticles, which could be used as probe in biosensors and bioassays. Rubpy2phen-5-NH2PF62 is has also been used to synthesize fluorescent silica nanoparticles with ultrabrightness and photostability by encapsulating it into silica nanoparticles by a modified reversed microemulsion method. Could be used for the determination of nucleotide base sequences.