Learn More
AdipoGen Rubpy2phen-ITC2
SDP
Catalog No. 502560139
Encompass_Preferred
Chemical. CAS 288399-07-5. Formula C33H23F12N7P2RuS. MW 940.65. Synthetic. Reactive probe containing fluorescent metal-ligand complex for determination of nucleotide base sequences.
Catalog No. 50-256-0139 Supplier AdipoGen Supplier No. CDXR0018M025
Chemical. CAS 288399-07-5. Formula C33H23F12N7P2RuS. MW 940.65. Synthetic. Reactive probe containing fluorescent metal-ligand complex for determination of nucleotide base sequences.