Learn More
Abcam Rubrato x in A, PP2A inhibitor, 250UG
Catalog No. 502705858
Encompass_Preferred
MW 520.5 Da, Purity >95%. Potent, selective, competitive PP2A inhibitor (Ki = 28.7 nM). Rubratoxin B analog. Induces caspase-dependent decrease in cell viability. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5858 Supplier Abcam Supplier No. AB144347250UG
MW 520.5 Da, Purity >95%. Potent, selective, competitive PP2A inhibitor (Ki = 28.7 nM). Rubratoxin B analog. Induces caspase-dependent decrease in cell viability. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement