Learn More
Abcam Rubrato x in A, PP2A inhibitor, 500UG
Catalog No. 502705857
Encompass_Preferred
MW 520.5 Da, Purity >95%. Potent, selective, competitive PP2A inhibitor (Ki = 28.7 nM). Rubratoxin B analog. Induces caspase-dependent decrease in cell viability. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5857 Supplier Abcam Supplier No. AB144347500UG
MW 520.5 Da, Purity >95%. Potent, selective, competitive PP2A inhibitor (Ki = 28.7 nM). Rubratoxin B analog. Induces caspase-dependent decrease in cell viability. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement