Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubratoxin B 98%, Thermo Scientific™

Catalog No. AC227390010
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227390010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227390010 Supplier Thermo Scientific Chemicals Supplier No. 227390010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21794-01-4
Molecular Formula C26H30O11
Molecular Weight (g/mol) 518.515
InChI Key ZJTBTDVZNGBSNG-WUCJZGDVSA-N
Synonym rubratoxin b, rubratoxinb, 4r*,5s* r* ,10s* s* s*-+-10-3,6-dihydro-6-oxo-2h-pyran-2-yl hydroxymethyl-5,9,10,11-tetrahydro-4-hydroxy-5-1-hydroxyheptyl-1h-cyclonona 1,2-c:5,6-c' difuran-1,3,6,8 4h-tetrone, rubratoxin b from penicillium rubrum
PubChem CID 133126381
SMILES CCCCCCC(C1C(C2=C(CC(CC3=C1C(=O)OC3=O)C(C4CC=CC(=O)O4)O)C(=O)OC2=O)O)O

Specifications

Quantity 1 mg
Formula Weight 518.52
Chemical Name or Material Rubratoxin B, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.