Learn More
MSC SPI Dial Indicator Contact Point, Cupped Ball, Ruby, #4-48 Thread, 0.7500 in., 1 ea.
Catalog No. 5000511563
Encompass
These precision contact points are designed for use with standard travel dial indicators. Featuring a cupped ball design, these components are engineered for durability and consistent measurement accuracy in demanding industrial applications.
  • Constructed with an extremely hard and smooth ruby measuring surface
  • Designed with a 4 mm ball size
  • Equipped with #4-48 standard threading
  • Features a 0.7500 in. point length
  • Compatible with all AGD Group 1, 2, 3 and 4 indicators
Catalog No. 50-005-11563 Supplier MSC Supplier No. 35915297
These precision contact points are designed for use with standard travel dial indicators. Featuring a cupped ball design, these components are engineered for durability and consistent measurement accuracy in demanding industrial applications.
  • Constructed with an extremely hard and smooth ruby measuring surface
  • Designed with a 4 mm ball size
  • Equipped with #4-48 standard threading
  • Features a 0.7500 in. point length
  • Compatible with all AGD Group 1, 2, 3 and 4 indicators