Learn More
Medchemexpress LLC HY-10617A 100mg , Rucaparib CAS:283173-50-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10617A100MG
Encompass_Preferred
HY-10617A 100mg Rucaparib CAS: 283173-50-2 Rucaparib (AG014699) is an orally active, potent inhibitor of PARP proteins (PARP-1, PARP-2 and PARP-3) with a Ki of 1.4 nM for PARP1. Rucaparib is a modest hexose-6-phosphate dehydrogenase (H6PD) inhibitor. Rucaparib has the potential for castration-resistant prostate cancer (CRPC) research[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9446