Learn More
Apexbio Technology LLC Rucaparib (AG-014699,PF-01367338) 459868-92-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061439
Encompass_Preferred
Rucaparib (AG-014699 PF-01367338 CAS 459868-92-9) is a small molecule inhibitor targeting poly(ADP-ribose) polymerase (PARP) a nuclear enzyme activated by DNA damage and central to the base excision repair pathway By blocking PARP activity rucaparib impairs DNA repair especially in cells with pre-existing DNA repair deficiencies such as those induced by genotoxic stress In prostate cancer research rucaparib induces radiosensitization notably in PTEN-deficient cells expressing ETS fusion proteins through inhibition of non-homologous end joining (NHEJ) It is thus relevant in studies investigating DNA damage response pathways cancer biology and therapeutic radiosensitization strategies
Catalog No. 50-000-61439 Supplier Apexbio Technology LLC Supplier No. A41565.1
Rucaparib (AG-014699 PF-01367338 CAS 459868-92-9) is a small molecule inhibitor targeting poly(ADP-ribose) polymerase (PARP) a nuclear enzyme activated by DNA damage and central to the base excision repair pathway By blocking PARP activity rucaparib impairs DNA repair especially in cells with pre-existing DNA repair deficiencies such as those induced by genotoxic stress In prostate cancer research rucaparib induces radiosensitization notably in PTEN-deficient cells expressing ETS fusion proteins through inhibition of non-homologous end joining (NHEJ) It is thus relevant in studies investigating DNA damage response pathways cancer biology and therapeutic radiosensitization strategies