Learn More
Medchemexpress LLC Rucaparib | 283173-50-2 | MFCD11977252 | 99.9% | 323.36 g·mol⁻¹ | C19H18FN3O | 1 MG
SDP
Catalog No. 5000241810
Encompass_Preferred
Rucaparib analytical reference standard provided as a characterized, high-purity material for research and analytical use. Supplied in a 1 mg pack, it is intended for method development, calibration, and compound identification in chromatographic and mass spectrometric workflows.
  • Analytical reference standard for rucaparib used in qualitative and quantitative assays.
  • High purity suitable for HPLC, GC, and mass spectrometry applications.
  • Packaged in milligram quantities for accurate calibration and method validation.
  • Chemically defined with documented molecular weight and formula for traceability.
  • For laboratory research use only; not for human consumption.
Catalog No. 50-002-41810 Supplier Medchemexpress LLC Supplier No. HY10617AR1MG
Rucaparib analytical reference standard provided as a characterized, high-purity material for research and analytical use. Supplied in a 1 mg pack, it is intended for method development, calibration, and compound identification in chromatographic and mass spectrometric workflows.
  • Analytical reference standard for rucaparib used in qualitative and quantitative assays.
  • High purity suitable for HPLC, GC, and mass spectrometry applications.
  • Packaged in milligram quantities for accurate calibration and method validation.
  • Chemically defined with documented molecular weight and formula for traceability.
  • For laboratory research use only; not for human consumption.