Learn More
Medchemexpress LLC HY-10617A 5mg Medchemexpress, Rucaparib CAS:283173-50-2 Purity:>98%
SDP
Catalog No. 502080688
Encompass_Preferred
Medchemexpress, HY-10617A 5mg Rucaparib CAS:283173-50-2 Rucaparib (AG014699) is an orally active and potent inhibitor of PARP with Ki of 1.4 nM for PARP1 in a cell-free assay. Rucaparib shows binding affinity to eight other PARP domains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0688 Supplier Medchemexpress LLC Supplier No. HY10617A5MG
Medchemexpress, HY-10617A 5mg Rucaparib CAS:283173-50-2 Rucaparib (AG014699) is an orally active and potent inhibitor of PARP with Ki of 1.4 nM for PARP1 in a cell-free assay. Rucaparib shows binding affinity to eight other PARP domains. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.