Learn More
Selleck Chemical LLC Rucaparib (AG-014699) phosphate 50mg 459868-92-9 PF-01367338
SDP
Catalog No. 508418
Encompass_Preferred
Rucaparib (AG-014699, PF-01367338) is an inhibitor of PARP with Ki of 1.4 nM for PARP1 in a cell-free assay, also showing binding affinity to eight other PARP domains. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-841-8 Supplier Selleck Chemical LLC Supplier No. S109850MG
Rucaparib (AG-014699, PF-01367338) is an inhibitor of PARP with Ki of 1.4 nM for PARP1 in a cell-free assay, also showing binding affinity to eight other PARP domains. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.